Assay Details
| Gene Symbol: LOC101697932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839947.3 | XP_004840004.1 | 3 - 4 | 512 | 72 | |
| XM_004839948.3 | XP_004840005.1 | 3 - 4 | 444 | 72 | ||
| XM_004839949.3 | XP_004840006.1 | 4 - 5 | 560 | 72 | ||
| XM_004839953.3 | XP_004840010.1 | 3 - 4 | 512 | 72 | ||
| XM_013070452.2 | XP_012925906.1 | 5 - 6 | 580 | 72 | ||
| XM_021254297.1 | XP_021109956.1 | 3 - 4 | 652 | 72 | ||
| XM_021254298.1 | XP_021109957.1 | 3 - 4 | 584 | 72 | ||
| XM_021254299.1 | XP_021109958.1 | 3 - 4 | 486 | 72 | ||
More Information
Set Membership: |
|

LOC101697932
Genomic Map