Assay Details
| Gene Symbol: LOC101697932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839947.3 | XP_004840004.1 | 5 - 6 | 734 | 121 | |
| XM_004839948.3 | XP_004840005.1 | 5 - 6 | 666 | 121 | ||
| XM_004839949.3 | XP_004840006.1 | 6 - 7 | 782 | 121 | ||
| XM_004839951.2 | XP_004840008.1 | 6 - 7 | 540 | 121 | ||
| XM_013070452.2 | XP_012925906.1 | 7 - 8 | 802 | 121 | ||
| XM_021254297.1 | XP_021109956.1 | 5 - 6 | 874 | 121 | ||
| XM_021254298.1 | XP_021109957.1 | 5 - 6 | 806 | 121 | ||
| XM_021254299.1 | XP_021109958.1 | 5 - 6 | 708 | 121 | ||
| XM_021254300.1 | XP_021109959.1 | 8 - 9 | 992 | 121 | ||
| XM_021254301.1 | XP_021109960.1 | 8 - 9 | 956 | 121 | ||
More Information
Set Membership: |
|

LOC101697932
Genomic Map