Assay Details
Gene Symbol: Nqo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847773.3 | XP_004847830.1 | 4 - 5 | 500 | 57 | |
XM_004847774.3 | XP_004847831.1 | 4 - 5 | 532 | 57 | ||
XM_004847775.3 | XP_004847832.1 | 4 - 5 | 467 | 57 | ||
XM_013073257.2 | XP_012928711.1 | 5 - 6 | 627 | 57 | ||
XM_013073258.2 | XP_012928712.1 | 5 - 6 | 584 | 57 | ||
XM_013073259.2 | XP_012928713.1 | 5 - 6 | 667 | 57 | ||
XM_021260744.1 | XP_021116403.1 | 4 - 5 | 762 | 57 | ||
XM_021260745.1 | XP_021116404.1 | 4 - 5 | 500 | 57 | ||
XM_021260746.1 | XP_021116405.1 | 4 - 5 | 619 | 57 | ||
XM_021260747.1 | XP_021116406.1 | 4 - 5 | 2112 | 57 | ||
XM_021260748.1 | XP_021116407.1 | 4 - 5 | 585 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
