Assay Details
| Gene Symbol: Fcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849001.2 | XP_004849058.1 | 5 - 6 | 365 | 90 | |
| XM_004849002.2 | XP_004849059.1 | 5 - 6 | 365 | 90 | ||
| XM_021261780.1 | XP_021117439.1 | 4 - 5 | 251 | 90 | ||
More Information
Set Membership: |
|

Fcn1
Genomic Map