Assay Details
| Gene Symbol: Tmem222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850700.3 | XP_004850757.1 | 3 - 4 | 311 | 58 | |
| XM_021263137.1 | XP_021118796.1 | 4 - 5 | 442 | 58 | ||
| XM_021263138.1 | XP_021118797.1 | 4 - 5 | 442 | 58 | ||
More Information
Set Membership: |
|

Tmem222
Genomic Map