Assay Details
| Gene Symbol: Acy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852215.2 | XP_004852272.1 | 6 - 7 | 834 | 56 | |
| XM_013074845.2 | XP_012930299.1 | 6 - 7 | 935 | 56 | ||
More Information
Set Membership: |
|

Acy3
Genomic Map