Assay Details
| Gene Symbol: Bnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852910.3 | XP_004852967.2 | 4 - 5 | 2271 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bnc1| Gene Symbol: Bnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852910.3 | XP_004852967.2 | 4 - 5 | 2271 | 74 | |
Set Membership: |
|