Assay Details
Gene Symbol: Il17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004854391.2 | XP_004854448.1 | 1 - 2 | 262 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004854391.2 transcripts.
Set Membership: |
![]() ![]() |
