Assay Details
| Gene Symbol: LOC101724434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870290.2 | XP_004870347.1 | 27 - 28 | 3512 | 69 | |
| XM_013067629.2 | XP_012923083.1 | 34 - 35 | 4928 | 69 | ||
| XM_021248594.1 | XP_021104253.1 | 33 - 34 | 4769 | 69 | ||
| XM_021248595.1 | XP_021104254.1 | 34 - 35 | 4925 | 69 | ||
| XM_021248596.1 | XP_021104255.1 | 33 - 34 | 4814 | 69 | ||
| XM_021248597.1 | XP_021104256.1 | 34 - 35 | 4928 | 69 | ||
| XM_021248598.1 | XP_021104257.1 | 32 - 33 | 4668 | 69 | ||
More Information
Set Membership: |
|

LOC101724434
Genomic Map