Assay Details
| Gene Symbol: LOC101724434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870290.2 | XP_004870347.1 | 9 - 10 | 1262 | 92 | |
| XM_013067629.2 | XP_012923083.1 | 16 - 17 | 2678 | 92 | ||
| XM_021248594.1 | XP_021104253.1 | 15 - 16 | 2519 | 92 | ||
| XM_021248595.1 | XP_021104254.1 | 16 - 17 | 2678 | 92 | ||
| XM_021248596.1 | XP_021104255.1 | 16 - 17 | 2678 | 92 | ||
| XM_021248597.1 | XP_021104256.1 | 16 - 17 | 2678 | 92 | ||
| XM_021248598.1 | XP_021104257.1 | 14 - 15 | 2418 | 92 | ||
| XM_021248599.1 | XP_021104258.1 | 16 - 17 | 2696 | 92 | ||
More Information
Set Membership: |
|

LOC101724434
Genomic Map