Assay Details
| Gene Symbol: LOC101697745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872540.2 | XP_004872597.2 | 1 - 2 | 425 | 76 | |
| XM_021250330.1 | XP_021105989.1 | 1 - 2 | 385 | 76 | ||
More Information
Set Membership: |
|

LOC101697745
Genomic Map