Assay Details
| Gene Symbol: LOC101697745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872540.2 | XP_004872597.2 | 4 - 5 | 812 | 74 | |
| XM_021250331.1 | XP_021105990.1 | 3 - 4 | 2323 | 74 | ||
More Information
Set Membership: |
|

LOC101697745
Genomic Map