Assay Details
| Gene Symbol: LOC101697745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872540.2 | XP_004872597.2 | 6 - 7 | 1061 | 92 | |
| XM_021250330.1 | XP_021105989.1 | 3 - 4 | 694 | 92 | ||
| XM_021250331.1 | XP_021105990.1 | 5 - 6 | 2572 | 92 | ||
More Information
Set Membership: |
|

LOC101697745
Genomic Map