Assay Details
| Gene Symbol: LOC101697745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872540.2 | XP_004872597.2 | 7 - 8 | 1301 | 120 | |
| XM_021250330.1 | XP_021105989.1 | 4 - 5 | 934 | 120 | ||
| XM_021250331.1 | XP_021105990.1 | 6 - 7 | 2812 | 120 | ||
More Information
Set Membership: |
|

LOC101697745
Genomic Map