Assay Details
| Gene Symbol: LOC106007526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875287.3 | XP_004875344.3 | 5 - 6 | 1030 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106007526| Gene Symbol: LOC106007526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875287.3 | XP_004875344.3 | 5 - 6 | 1030 | 96 | |
Set Membership: |
|