Assay Details
| Gene Symbol: Tbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067921.2 | XP_012923375.1 | 2 - 3 | 891 | 60 | |
| XM_021249232.1 | XP_021104891.1 | 2 - 3 | 891 | 60 | ||
| XM_021249233.1 | XP_021104892.1 | 2 - 3 | 891 | 60 | ||
| XM_021249234.1 | XP_021104893.1 | 2 - 3 | 890 | 60 | ||
More Information
Set Membership: |
|

Tbpl2
Genomic Map