Assay Details
| Gene Symbol: Nptxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072479.2 | XP_012927933.2 | 2 - 3 | 981 | 96 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nptxr| Gene Symbol: Nptxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072479.2 | XP_012927933.2 | 2 - 3 | 981 | 96 | |