Assay Details
| Gene Symbol: Myrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072884.2 | XP_012928338.2 | 7 - 8 | 877 | 59 | |
| XM_021258056.1 | XP_021113715.1 | 7 - 8 | 878 | 59 | ||
| XM_021258058.1 | XP_021113717.1 | 7 - 8 | 879 | 59 | ||
| XM_021258063.1 | XP_021113722.1 | 7 - 8 | 878 | 59 | ||
| XM_021258069.1 | XP_021113728.1 | 7 - 8 | 878 | 59 | ||
More Information
Set Membership: |
|

Myrip
Genomic Map