Assay Details
| Gene Symbol: Znf570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077432.2 | XP_012932886.1 | 2 - 3 | 278 | 91 | |
| XM_013077435.2 | XP_012932889.1 | 2 - 3 | 299 | 91 | ||
| XM_013077436.2 | XP_012932890.1 | 2 - 3 | 288 | 91 | ||
| XM_021239494.1 | XP_021095153.1 | 2 - 3 | 385 | 91 | ||
| XM_021239495.1 | XP_021095154.1 | 2 - 3 | 112 | 91 | ||

Znf570
Genomic Map