Assay Details
| Gene Symbol: Znf570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077432.2 | XP_012932886.1 | 3 - 4 | 400 | 96 | |
| XM_013077435.2 | XP_012932889.1 | 3 - 4 | 421 | 96 | ||
| XM_013077436.2 | XP_012932890.1 | 3 - 4 | 410 | 96 | ||
| XM_021239494.1 | XP_021095153.1 | 3 - 4 | 507 | 96 | ||
| XM_021239495.1 | XP_021095154.1 | 3 - 4 | 234 | 96 | ||
| XM_021239496.1 | XP_021095155.1 | 4 - 5 | 586 | 96 | ||

Znf570
Genomic Map