Assay Details
| Gene Symbol: Znf570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077432.2 | XP_012932886.1 | 4 - 5 | 498 | 73 | |
| XM_013077435.2 | XP_012932889.1 | 4 - 5 | 519 | 73 | ||
| XM_013077436.2 | XP_012932890.1 | 4 - 5 | 508 | 73 | ||
| XM_021239494.1 | XP_021095153.1 | 4 - 5 | 605 | 73 | ||
| XM_021239495.1 | XP_021095154.1 | 4 - 5 | 332 | 73 | ||
| XM_021239496.1 | XP_021095155.1 | 5 - 6 | 684 | 73 | ||
| XM_021239497.1 | XP_021095156.1 | 2 - 3 | 183 | 73 | ||
More Information
Set Membership: |
|

Znf570
Genomic Map