Assay Details
| Gene Symbol: Syce1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256688.1 | XP_021112347.1 | 3 - 4 | 1950 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syce1l| Gene Symbol: Syce1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256688.1 | XP_021112347.1 | 3 - 4 | 1950 | 64 | |
Set Membership: |
|