Assay Details
| Gene Symbol: LOC110348804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257028.1 | XP_021112687.1 | 5 - 6 | 748 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110348804| Gene Symbol: LOC110348804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257028.1 | XP_021112687.1 | 5 - 6 | 748 | 74 | |
Set Membership: |
|