Assay Details
| Gene Symbol: LOC110348959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257766.1 | XP_021113425.1 | 1 - 2 | 1132 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110348959| Gene Symbol: LOC110348959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257766.1 | XP_021113425.1 | 1 - 2 | 1132 | 74 | |
Set Membership: |
|