Assay Details
| Gene Symbol: Lsmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835141.3 | XP_004835198.2 | 1 - 2 | 346 | 83 | |
| XM_013070293.2 | XP_012925747.1 | 1 - 2 | 346 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004835141.3, XM_013070293.2 transcripts.
Set Membership: |
|

Lsmem2
Genomic Map