Assay Details
| Gene Symbol: Lsmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835141.3 | XP_004835198.2 | 2 - 3 | 458 | 70 | |
| XM_013070293.2 | XP_012925747.1 | 2 - 3 | 458 | 70 | ||
More Information
Set Membership: |
|

Lsmem2
Genomic Map