Assay Details
| Gene Symbol: Atp5f1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840969.2 | XP_004841026.1 | 1 - 2 | 179 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004840969.2 transcripts.
Set Membership: |
|

Atp5f1e
Genomic Map