Assay Details
| Gene Symbol: Atp5f1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840969.2 | XP_004841026.1 | 2 - 3 | 316 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp5f1e| Gene Symbol: Atp5f1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840969.2 | XP_004841026.1 | 2 - 3 | 316 | 100 | |
Set Membership: |
|