Assay Details
| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841430.3 | XP_004841487.1 | 7 - 8 | 710 | 82 | |
| XM_004841431.3 | XP_004841488.1 | 5 - 6 | 542 | 82 | ||
More Information
Set Membership: |
|

Acadm
Genomic Map