Assay Details
| Gene Symbol: LOC101720625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843060.3 | XP_004843117.1 | 1 - 2 | 521 | 62 | |
| XM_013071573.2 | XP_012927027.1 | 1 - 2 | 521 | 62 | ||
More Information
Set Membership: |
|

LOC101720625
Genomic Map