Assay Details
| Gene Symbol: LOC101720625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843060.3 | XP_004843117.1 | 9 - 10 | 1484 | 101 | |
| XM_004843061.2 | XP_004843118.1 | 8 - 9 | 902 | 101 | ||
| XM_013071573.2 | XP_012927027.1 | 9 - 10 | 1484 | 101 | ||
More Information
Set Membership: |
|

LOC101720625
Genomic Map