Assay Details
| Gene Symbol: Adgrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843154.3 | XP_004843211.1 | 3 - 4 | 2540 | 82 | |
| XM_013071612.2 | XP_012927066.1 | 3 - 4 | 2542 | 82 | ||
More Information
Set Membership: |
|

Adgrg3
Genomic Map