Assay Details
| Gene Symbol: Adgrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843154.3 | XP_004843211.1 | 4 - 5 | 2687 | 81 | |
| XM_013071612.2 | XP_012927066.1 | 4 - 5 | 2689 | 81 | ||
More Information
Set Membership: |
|

Adgrg3
Genomic Map