Assay Details
| Gene Symbol: Sp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857693.3 | XP_004857750.1 | 3 - 4 | 2143 | 68 | |
| XM_021238557.1 | XP_021094216.1 | 2 - 3 | 1441 | 68 | ||
| XM_021238558.1 | XP_021094217.1 | 3 - 4 | 2145 | 68 | ||
| XM_021238559.1 | XP_021094218.1 | 4 - 5 | 2270 | 68 | ||
More Information
Set Membership: |
|

Sp3
Genomic Map