Assay Details
| Gene Symbol: Actn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859057.2 | XP_004859114.1 | 14 - 15 | 1805 | 59 | |
| XM_004859058.3 | XP_004859115.1 | 14 - 15 | 1806 | 59 | ||
| XM_004859059.3 | XP_004859116.1 | 14 - 15 | 1804 | 59 | ||
| XM_004859060.3 | XP_004859117.1 | 14 - 15 | 1805 | 59 | ||
| XM_004859061.3 | XP_004859118.1 | 14 - 15 | 1806 | 59 | ||
More Information
Set Membership: |
|

Actn4
Genomic Map