Assay Details
| Gene Symbol: Sdf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863882.3 | XP_004863939.1 | 3 - 4 | 703 | 55 | |
| XM_004863884.3 | XP_004863941.1 | 3 - 4 | 715 | 55 | ||
| XM_021243492.1 | XP_021099151.1 | 3 - 4 | 702 | 55 | ||
| XM_021243493.1 | XP_021099152.1 | 3 - 4 | 700 | 55 | ||
More Information
Set Membership: |
|

Sdf4
Genomic Map