Assay Details
| Gene Symbol: Tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873306.2 | XP_004873363.1 | 8 - 9 | 829 | 73 | |
| XM_004873308.2 | XP_004873365.1 | 7 - 8 | 728 | 73 | ||
| XM_004873311.3 | XP_004873368.1 | 7 - 8 | 667 | 73 | ||
| XM_021251112.1 | XP_021106771.1 | 7 - 8 | 682 | 73 | ||
More Information
Set Membership: |
|

Tpm4
Genomic Map