Assay Details
| Gene Symbol: Rfxank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873420.3 | XP_004873477.1 | 4 - 5 | 583 | 61 | |
| XM_004873422.3 | XP_004873479.1 | 4 - 5 | 569 | 61 | ||
| XM_013068724.2 | XP_012924178.1 | 4 - 5 | 702 | 61 | ||
| XM_021251204.1 | XP_021106863.1 | 4 - 5 | 576 | 61 | ||
More Information
Set Membership: |
|

Rfxank
Genomic Map