Assay Details
| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073884.2 | XP_012929338.1 | 3 - 4 | 456 | 133 | |
| XM_013073885.2 | XP_012929339.1 | 3 - 4 | 447 | 133 | ||
| XM_013073886.2 | XP_012929340.1 | 3 - 4 | 513 | 133 | ||
| XM_013073887.2 | XP_012929341.1 | 3 - 4 | 479 | 133 | ||
| XM_013073888.2 | XP_012929342.1 | 3 - 4 | 561 | 133 | ||
| XM_013073890.2 | XP_012929344.1 | 4 - 5 | 522 | 133 | ||
More Information
Set Membership: |
|

Zfp37
Genomic Map