Assay Details
Gene Symbol: LOC101720600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021239518.1 | XP_021095177.1 | 1 - 2 | 368 | 59 | |
XM_021239521.1 | XP_021095180.1 | 1 - 2 | 376 | 59 | ||
XM_021239522.1 | XP_021095181.1 | 2 - 3 | 218 | 59 | ||
XM_021239534.1 | XP_021095193.1 | 1 - 2 | 378 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021239522.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
