Assay Details
| Gene Symbol: Rassf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021241425.1 | XP_021097084.1 | 4 - 5 | 657 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rassf3| Gene Symbol: Rassf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021241425.1 | XP_021097084.1 | 4 - 5 | 657 | 69 | |
Set Membership: |
|