Assay Details
| Gene Symbol: Npffr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259543.1 | XP_021115202.1 | 1 - 2 | 4449 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npffr1| Gene Symbol: Npffr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259543.1 | XP_021115202.1 | 1 - 2 | 4449 | 67 | |
Set Membership: |
|