Assay Details
| Gene Symbol: Npffr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259543.1 | XP_021115202.1 | 2 - 3 | 4547 | 72 | |
| XM_021259544.1 | XP_021115203.1 | 3 - 4 | 452 | 72 | ||
More Information
Set Membership: |
|

Npffr1
Genomic Map