Assay Details
| Gene Symbol: Tesmin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021264474.1 | XP_021120133.1 | 4 - 5 | 877 | 76 | |
| XM_021264475.1 | XP_021120134.1 | 4 - 5 | 956 | 76 | ||
| XM_021264476.1 | XP_021120135.1 | 4 - 5 | 877 | 76 | ||
| XM_021264477.1 | XP_021120136.1 | 4 - 5 | 877 | 76 | ||
More Information
Set Membership: |
|

Tesmin
Genomic Map