Assay Details
| Gene Symbol: CUNH3orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834021.2 | XP_004834078.2 | 3 - 4 | 958 | 111 | |
| XM_013070052.2 | XP_012925506.1 | 3 - 4 | 975 | 111 | ||
| XM_021261392.1 | XP_021117051.1 | 2 - 3 | 634 | 111 | ||
More Information
Set Membership: |
|

CUNH3orf62
Genomic Map