Assay Details
| Gene Symbol: Gmnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834779.2 | XP_004834836.1 | 3 - 4 | 325 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmnc| Gene Symbol: Gmnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834779.2 | XP_004834836.1 | 3 - 4 | 325 | 71 | |
Set Membership: |
|