Assay Details
| Gene Symbol: Pank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840759.3 | XP_004840816.1 | 2 - 3 | 740 | 78 | |
| XM_021255010.1 | XP_021110669.1 | 2 - 3 | 738 | 78 | ||
| XM_021255011.1 | XP_021110670.1 | 2 - 3 | 818 | 78 | ||
More Information
Set Membership: |
|

Pank2
Genomic Map