Assay Details
| Gene Symbol: Setd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843239.2 | XP_004843296.1 | 7 - 8 | 1169 | 69 | |
| XM_021256969.1 | XP_021112628.1 | 7 - 8 | 1106 | 69 | ||
More Information
Set Membership: |
|

Setd6
Genomic Map