Assay Details
| Gene Symbol: Ckap2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844528.3 | XP_004844585.1 | 6 - 7 | 1847 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ckap2l| Gene Symbol: Ckap2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844528.3 | XP_004844585.1 | 6 - 7 | 1847 | 65 | |
Set Membership: |
|