Assay Details
| Gene Symbol: Pbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846874.3 | XP_004846931.1 | 4 - 5 | 796 | 59 | |
| XM_004846875.3 | XP_004846932.1 | 4 - 5 | 977 | 59 | ||
More Information
Set Membership: |
|

Pbx2
Genomic Map